siRNA Details
Gene Symbol: NPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037738.2 | NP_001032827.1 | 4, 5 | 602 | ||
NM_002520.6 | NP_002511.1 | 4, 5 | 602 | |||
NM_199185.3 | NP_954654.1 | 4, 5 | 602 | |||
XM_005265920.3 | XP_005265977.1 | 4, 5 | 658 | |||
XM_011534564.1 | XP_011532866.1 | 3, 4 | 422 | |||
GenBank mRNA | AB042278.1 | - | 4, 5 | 357 | ||
AB451236.1 | - | 4, 5 | 357 | |||
AB451361.1 | - | 4, 5 | 357 | |||
AK000472.1 | - | 2, 3 | 165 | |||
AK290652.1 | - | 4, 5 | 465 | |||
AY347529.1 | - | 5, 6 | 492 | |||
AY740634.1 | - | 4, 5 | 429 | |||
AY740635.1 | - | 4, 5 | 429 | |||
AY740636.1 | - | 4, 5 | 429 | |||
AY740637.1 | - | 4, 5 | 429 | |||
AY740638.1 | - | 4, 5 | 429 | |||
AY740639.1 | - | 4, 5 | 429 | |||
AY740640.1 | - | 4, 5 | 429 | |||
BC002398.2 | - | 4, 5 | 444 | 2821577 | ||
BC003670.1 | - | 1 | 159 | |||
BC008495.1 | - | 4, 5 | 454 | 4276604 | ||
BC009623.2 | - | 4, 5 | 483 | 3877633 | ||
BC012566.1 | - | 4, 5 | 457 | 4097025 | ||
BC014349.1 | - | 4, 5 | 453 | 3511037 | ||
BC016716.1 | - | 4, 5 | 456 | 4068167 | ||
BC016768.1 | - | 4, 5 | 456 | 4081364 | ||
BC016824.1 | - | 4, 5 | 456 | 4106286 | ||
BC020467.1 | - | 4, 5 | 443 | |||
BC021668.1 | - | 4, 5 | 457 | 3930893 | ||
BC021983.1 | - | 4, 5 | 455 | 4106023 | ||
BC050628.1 | - | 4, 5 | 443 | 5575414 | ||
BC107754.1 | - | 4, 5 | 458 | 6736004 | ||
BT007011.1 | - | 4, 5 | 357 | |||
DQ303464.1 | - | 4, 5 | 453 | |||
EF429251.1 | - | 4, 5 | 357 | |||
KU178236.1 | - | 4, 5 | 357 | |||
KU178237.1 | - | 4, 5 | 357 | |||
M23613.1 | - | 4, 5 | 450 | |||
M26697.1 | - | 4, 5 | 447 | |||
M28699.1 | - | 4, 5 | 444 | |||
X16934.1 | - | 4, 5 | 317 |
Gene Symbol: BLNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011539728.2 | XP_011538030.1 | 17 | 3534 | ||
XM_017016159.1 | XP_016871648.1 | 16 | 3466 |
Gene Symbol: FANCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029356.1 | XP_016884845.1 | 11 | 3734 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
