siRNA Details
Gene Symbol: FRMPD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033172.1 | 5 | 814 | |||
GenBank mRNA | AK294650.1 | - | 5 | 769 | ||
AK313659.1 | - | 5 | 771 | |||
AK315971.1 | - | 5 | 814 | |||
BC071635.1 | - | 5 | 726 | |||
BC072395.1 | - | 5 | 740 | |||
BC110798.1 | - | 5 | 737 | |||
BC137431.1 | - | 5 | 701 | 9021429 | ||
BC137434.1 | - | 5 | 701 | 9021432 | ||
BC137439.1 | - | 5 | 699 | 9021437 | ||
BC137440.1 | - | 5 | 701 | 9021438 | ||
BC144635.1 | - | 5 | 701 | 9053166 |
Gene Symbol: FRMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018071.3 | NP_001018081.3 | 28 | 3892 | ||
NM_001042512.2 | NP_001035977.2 | 5 | 760 | |||
NM_001318191.1 | NP_001305120.1 | 26 | 3533 | |||
XM_017015744.1 | XP_016871233.1 | 5 | 576 | |||
GenBank mRNA | AK123038.1 | - | 26 | 3533 | ||
AY491519.1 | - | 5 | 759 | |||
BC031614.1 | - | 7 | 755 | |||
BC073954.1 | - | 5 | 741 | |||
BC144636.1 | - | 5 | 701 | 9053167 | ||
BC144639.1 | - | 5 | 701 | 9053170 | ||
CR749241.1 | - | 28 | 3864 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
