siRNA Details
Gene Symbol: FRMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018071.3 | NP_001018081.3 | 26 | 3656 | ||
NM_001042512.2 | NP_001035977.2 | 3 | 524 | |||
NM_001318191.1 | NP_001305120.1 | 24 | 3297 | |||
XM_017015744.1 | XP_016871233.1 | 3 | 340 | |||
GenBank mRNA | AK123038.1 | - | 24 | 3297 | ||
AY491519.1 | - | 3 | 523 | |||
BC031614.1 | - | 5 | 519 | |||
BC073954.1 | - | 3 | 505 | |||
BC144636.1 | - | 3 | 465 | 9053167 | ||
BC144639.1 | - | 3 | 465 | 9053170 | ||
CR749241.1 | - | 26 | 3628 |
Gene Symbol: FRMPD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033172.1 | 3 | 578 | |||
GenBank mRNA | AK294650.1 | - | 3 | 533 | ||
AK313659.1 | - | 3 | 535 | |||
AK315971.1 | - | 3 | 578 | |||
BC071635.1 | - | 3 | 490 | |||
BC072395.1 | - | 3 | 504 | |||
BC110798.1 | - | 3 | 501 | |||
BC137431.1 | - | 3 | 465 | 9021429 | ||
BC137434.1 | - | 3 | 465 | 9021432 | ||
BC137439.1 | - | 3 | 463 | 9021437 | ||
BC137440.1 | - | 3 | 465 | 9021438 | ||
BC144635.1 | - | 3 | 465 | 9053166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
