siRNA Details
Gene Symbol: FRMPD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033172.1 | 5 | 1113 | |||
GenBank mRNA | AK294650.1 | - | 5 | 1068 | ||
AK313659.1 | - | 5 | 1070 | |||
AK315971.1 | - | 5 | 1113 | |||
BC071635.1 | - | 5 | 1025 | |||
BC072395.1 | - | 5 | 1039 | |||
BC110798.1 | - | 5 | 1036 | |||
BC137431.1 | - | 5 | 1000 | 9021429 | ||
BC137434.1 | - | 5 | 1000 | 9021432 | ||
BC137439.1 | - | 5 | 998 | 9021437 | ||
BC137440.1 | - | 5 | 1000 | 9021438 | ||
BC144635.1 | - | 5 | 1000 | 9053166 |
Gene Symbol: FRMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018071.3 | NP_001018081.3 | 28 | 4191 | ||
NM_001042512.2 | NP_001035977.2 | 5 | 1059 | |||
NM_001318191.1 | NP_001305120.1 | 26 | 3832 | |||
XM_017015744.1 | XP_016871233.1 | 5 | 875 | |||
GenBank mRNA | AK123038.1 | - | 26 | 3832 | ||
AY491519.1 | - | 5 | 1058 | |||
BC031614.1 | - | 7 | 1054 | |||
BC073954.1 | - | 5 | 1040 | |||
BC144636.1 | - | 5 | 1000 | 9053167 | ||
BC144639.1 | - | 5 | 1000 | 9053170 | ||
CR749241.1 | - | 28 | 4163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
