siRNA Details
Gene Symbol: POM121L8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024583.1 | 7 | 4748 | |||
GenBank mRNA | AL117485.1 | - | 5 | 2699 | ||
BC050331.1 | - | 2 | 932 | |||
BC068032.1 | - | 2 | 793 | |||
BC090873.1 | - | 2 | 653 |
Gene Symbol: LOC102725072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135922.1 | 6 | 3687 | |||
GenBank mRNA | BC051721.1 | - | 6 | 3687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
