siRNA Details
Gene Symbol: FAM138E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026819.1 | 2, 3 | 565 |
Gene Symbol: FAM138A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026818.1 | 2, 3 | 565 | |||
GenBank mRNA | AY341950.1 | - | 2, 3 | 561 |
Gene Symbol: FAM138C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026822.1 | 2, 3 | 565 | |||
GenBank mRNA | AY341953.1 | - | 2, 3 | 561 |
Gene Symbol: FAM138F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026820.1 | 2, 3 | 565 | |||
GenBank mRNA | AY341952.1 | - | 2, 3 | 561 | ||
BC137207.1 | - | 2, 3 | 397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
