siRNA Details
Gene Symbol: CKS1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001826.2 | NP_001817.1 | 2, 3 | 285 | ||
NR_024163.1 | 2, 3 | 393 | ||||
GenBank mRNA | AF274941.1 | - | 1 | 207 | ||
AF274943.1 | - | 2, 3 | 213 | |||
AF279897.1 | - | 2, 3 | 207 | |||
AK307066.1 | - | 2, 3 | 341 | |||
AK311769.1 | - | 2, 3 | 246 | |||
BC007751.2 | - | 2, 3 | 228 | 4110344 | ||
BC015629.2 | - | 2, 3 | 207 | 3162564 | ||
BC070319.1 | - | 2, 3 | 246 | 4738841 | ||
BC070320.1 | - | 2, 3 | 246 | 6494754 | ||
BQ883992.1 | - | 2, 3 | 224 | |||
BT007196.1 | - | 2, 3 | 181 | |||
CB161621.1 | - | 2, 3 | 344 | |||
CN347951.1 | - | 2, 3 | 287 | |||
X54941.1 | - | 2, 3 | 190 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_016904, BC076579, AK011314, AK010414, AK011782, and others.
Gene Ontology Categories:
