siRNA Details
Gene Symbol: IL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310053.1 | - | 3 | 1235 |
Gene Symbol: ANGPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK225698.1 | - | 8 | 2309 |
Gene Symbol: EMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK296184.1 | - | 2 | 747 |
Gene Symbol: NSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC144629.1 | - | 3 | 341 |
Gene Symbol: PTGER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC118578.1 | - | 5 | 1386 | 40069637 | |
BC118659.1 | - | 4 | 1335 | 40069636 | ||
U13216.1 | - | 6 | 1487 |
Gene Symbol: RAB22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020673.2 | NP_065724.1 | 7 | 6085 | ||
GenBank mRNA | AK054767.1 | - | 1 | 295 | ||
BC042545.1 | - | 1 | 156 |
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122742.1 | - | 1 | 727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
