siRNA Details
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273603.4 | XP_005273660.1 | 7 | 1611 | ||
XM_005273604.4 | XP_005273661.1 | 6 | 1530 | |||
XM_005273605.4 | XP_005273662.1 | 6 | 1431 | |||
XM_005273607.2 | XP_005273664.1 | 6 | 1469 | |||
XM_006716387.3 | XP_006716450.1 | 7 | 1632 | |||
XM_011544618.2 | XP_011542920.1 | 8 | 1683 | |||
XM_011544619.2 | XP_011542921.1 | 7 | 1650 | |||
XM_011544622.2 | XP_011542924.1 | 7 | 1552 | |||
XM_017013740.1 | XP_016869229.1 | 6 | 1613 | |||
XM_017013741.1 | XP_016869230.1 | 5 | 1550 | |||
XM_017013742.1 | XP_016869231.1 | 5 | 1532 |
Gene Symbol: ALG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ002699.1 | - | 1 | 463 | ||
CD366777.1 | - | 1 | 269 |
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289029.1 | NP_001275958.1 | 6 | 1381 | ||
NM_201400.3 | NP_958802.1 | 8 | 1564 | |||
NM_201598.3 | NP_963892.1 | 7 | 1462 | |||
XM_005255157.4 | XP_005255214.1 | 7 | 1456 | |||
XM_005255158.4 | XP_005255215.1 | 9 | 1610 | |||
XM_011522404.2 | XP_011520706.1 | 8 | 1529 | |||
GenBank mRNA | BC010084.2 | - | 8 | 1484 | 2822323 | |
BI491350.1 | - | 1 | 480 | |||
BQ215173.1 | - | 2 | 628 |
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 7 | 1581 | ||
NR_003494.1 | 6 | 1315 | ||||
XM_006716256.3 | XP_006716319.1 | 8 | 1676 | |||
XM_006716257.3 | XP_006716320.1 | 7 | 1746 | |||
XM_006716260.3 | XP_006716323.1 | 6 | 1490 | |||
XM_006716262.1 | XP_006716325.1 | 6 | 1333 | |||
XM_011543842.2 | XP_011542144.1 | 6 | 1755 | |||
XM_011543843.2 | XP_011542145.1 | 7 | 1694 | |||
XM_017013916.1 | XP_016869405.1 | 7 | 1722 | |||
XM_017013917.1 | XP_016869406.1 | 6 | 1643 | |||
XM_017013918.1 | XP_016869407.1 | 6 | 1615 | |||
XM_017013919.1 | XP_016869408.1 | 5 | 1204 | |||
GenBank mRNA | AK098138.1 | - | 7 | 1178 | ||
BC062615.1 | - | 7 | 1255 | |||
BC085016.1 | - | 8 | 1284 | |||
BC093808.1 | - | 1 | 130 | |||
BC093810.1 | - | 1 | 130 | |||
BC098427.1 | - | 7 | 1261 |
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 5 | 1267 | ||
NM_018172.2 | NP_060642.2 | 5 | 1288 | |||
NM_152563.2 | NP_689776.1 | 4 | 1186 | |||
GenBank mRNA | AK001523.1 | - | 5 | 1262 | ||
AK130709.1 | - | 4 | 1183 | |||
AK311428.1 | - | 5 | 1244 | |||
AK311518.1 | - | 5 | 1267 | |||
BC032519.1 | - | 4 | 1186 | 5494088 |
Gene Symbol: FAM86FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024254.1 | 5 | 1175 |
Gene Symbol: FAM86HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024252.1 | 5 | 1286 | |||
GenBank mRNA | AK124448.1 | - | 3 | 1054 |
Gene Symbol: FAM86JP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024250.1 | 5 | 1094 | |||
NR_024251.1 | 5 | 1192 | ||||
GenBank mRNA | BC032772.1 | - | 5 | 1080 | ||
BC041080.1 | - | 5 | 1163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
