siRNA Details
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 7 | 1650 | ||
NR_003494.1 | 6 | 1384 | ||||
XM_006716256.3 | XP_006716319.1 | 8 | 1745 | |||
XM_006716257.3 | XP_006716320.1 | 7 | 1815 | |||
XM_006716260.3 | XP_006716323.1 | 6 | 1559 | |||
XM_006716262.1 | XP_006716325.1 | 6 | 1402 | |||
XM_011543842.2 | XP_011542144.1 | 6 | 1824 | |||
XM_011543843.2 | XP_011542145.1 | 7 | 1763 | |||
XM_017013916.1 | XP_016869405.1 | 7 | 1791 | |||
XM_017013917.1 | XP_016869406.1 | 6 | 1712 | |||
XM_017013918.1 | XP_016869407.1 | 6 | 1684 | |||
XM_017013919.1 | XP_016869408.1 | 5 | 1273 | |||
GenBank mRNA | AK098138.1 | - | 7 | 1247 | ||
BC062615.1 | - | 7 | 1324 | |||
BC085016.1 | - | 8 | 1353 | |||
BC093808.1 | - | 1 | 199 | |||
BC093810.1 | - | 1 | 199 | |||
BC098427.1 | - | 7 | 1330 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273603.4 | XP_005273660.1 | 7 | 1680 | ||
XM_005273604.4 | XP_005273661.1 | 6 | 1599 | |||
XM_005273605.4 | XP_005273662.1 | 6 | 1500 | |||
XM_005273607.2 | XP_005273664.1 | 6 | 1538 | |||
XM_006716387.3 | XP_006716450.1 | 7 | 1701 | |||
XM_011544618.2 | XP_011542920.1 | 8 | 1752 | |||
XM_011544619.2 | XP_011542921.1 | 7 | 1719 | |||
XM_011544622.2 | XP_011542924.1 | 7 | 1621 | |||
XM_017013740.1 | XP_016869229.1 | 6 | 1682 | |||
XM_017013741.1 | XP_016869230.1 | 5 | 1619 | |||
XM_017013742.1 | XP_016869231.1 | 5 | 1601 |
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 5 | 1336 | ||
NM_018172.2 | NP_060642.2 | 5 | 1357 | |||
NM_152563.2 | NP_689776.1 | 4 | 1255 | |||
GenBank mRNA | AK001523.1 | - | 5 | 1331 | ||
AK130709.1 | - | 4 | 1252 | |||
AK311428.1 | - | 5 | 1313 | |||
AK311518.1 | - | 5 | 1336 | |||
BC032519.1 | - | 4 | 1255 | 5494088 |
Gene Symbol: FAM86DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024241.1 | 8 | 1742 | |||
GenBank mRNA | BC043342.2 | - | 7 | 1466 |
Gene Symbol: FAM86FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024254.1 | 5 | 1247 |
Gene Symbol: FAM86HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024252.1 | 5 | 1358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
