siRNA Details
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273603.4 | XP_005273660.1 | 7 | 1657 | ||
XM_005273604.4 | XP_005273661.1 | 6 | 1576 | |||
XM_005273605.4 | XP_005273662.1 | 6 | 1477 | |||
XM_005273607.2 | XP_005273664.1 | 6 | 1515 | |||
XM_006716387.3 | XP_006716450.1 | 7 | 1678 | |||
XM_011544618.2 | XP_011542920.1 | 8 | 1729 | |||
XM_011544619.2 | XP_011542921.1 | 7 | 1696 | |||
XM_011544622.2 | XP_011542924.1 | 7 | 1598 | |||
XM_017013740.1 | XP_016869229.1 | 6 | 1659 | |||
XM_017013741.1 | XP_016869230.1 | 5 | 1596 | |||
XM_017013742.1 | XP_016869231.1 | 5 | 1578 |
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 7 | 1627 | ||
NR_003494.1 | 6 | 1361 | ||||
XM_006716256.3 | XP_006716319.1 | 8 | 1722 | |||
XM_006716257.3 | XP_006716320.1 | 7 | 1792 | |||
XM_006716260.3 | XP_006716323.1 | 6 | 1536 | |||
XM_006716262.1 | XP_006716325.1 | 6 | 1379 | |||
XM_011543842.2 | XP_011542144.1 | 6 | 1801 | |||
XM_011543843.2 | XP_011542145.1 | 7 | 1740 | |||
XM_017013916.1 | XP_016869405.1 | 7 | 1768 | |||
XM_017013917.1 | XP_016869406.1 | 6 | 1689 | |||
XM_017013918.1 | XP_016869407.1 | 6 | 1661 | |||
XM_017013919.1 | XP_016869408.1 | 5 | 1250 | |||
GenBank mRNA | AK098138.1 | - | 7 | 1224 | ||
BC062615.1 | - | 7 | 1301 | |||
BC085016.1 | - | 8 | 1330 | |||
BC093808.1 | - | 1 | 176 | |||
BC093810.1 | - | 1 | 176 | |||
BC098427.1 | - | 7 | 1307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
