siRNA Details
Gene Symbol: NUDT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018159.3 | NP_060629.2 | 1 | 519 | ||
GenBank mRNA | AK001490.1 | - | 1 | 406 | ||
BC009942.2 | - | 1 | 473 | 4303422 | ||
BX419815.2 | - | 1 | 505 |
Gene Symbol: NUDT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304963.1 | NP_001291892.1 | 1 | 513 | ||
NM_153183.3 | NP_694853.1 | 2 | 576 | |||
GenBank mRNA | AF469196.1 | - | 2 | 578 | ||
AK291952.1 | - | 1 | 493 | |||
BC049383.1 | - | 1 | 434 | 5725355 | ||
BC050700.1 | - | 1 | 492 | 5723622 | ||
CN399689.1 | - | 1 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
