siRNA Details
Gene Symbol: ADAMTS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014272.4 | NP_055087.2 | 2 | 638 | ||
XM_005254137.3 | XP_005254194.1 | 2 | 639 | |||
GenBank mRNA | AB209497.1 | - | 1 | 292 | ||
AF140675.1 | - | 2 | 407 | |||
AY327122.1 | - | 2 | 405 |
Gene Symbol: ADAMTS7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045529.3 | 2 | 347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
