siRNA Details
Gene Symbol: CPAMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015692.2 | NP_056507.2 | 8 | 744 | ||
XM_011527917.1 | XP_011526219.1 | 8 | 711 | |||
XM_011527918.1 | XP_011526220.1 | 8 | 747 | |||
XM_011527919.1 | XP_011526221.1 | 8 | 747 | |||
XM_011527920.2 | XP_011526222.1 | 8 | 747 | |||
XM_011527921.2 | XP_011526223.1 | 8 | 747 | |||
XM_011527922.1 | XP_011526224.1 | 8 | 747 | |||
XM_011527923.1 | XP_011526225.1 | 8 | 747 | |||
XM_011527924.2 | XP_011526226.1 | 8 | 747 | |||
XM_017026594.1 | XP_016882083.1 | 8 | 747 | |||
GenBank mRNA | AB033109.1 | - | 8 | 569 | ||
AY101765.1 | - | 8 | 588 | |||
BC067854.1 | - | 8 | 744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
