siRNA Details
Gene Symbol: SEC11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209437.1 | - | 1 | 851 |
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336879.1 | - | 1 | 664 | ||
AF336883.1 | - | 1 | 689 | |||
AF469044.1 | - | 1 | 689 |
Gene Symbol: LOC400002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135047.1 | 1 | 411 | |||
GenBank mRNA | AK128230.1 | - | 1 | 412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
