siRNA Details
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 4491 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 3557 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 4358 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 4045 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 4620 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 4884 | |||
NM_003879.5 | NP_003870.4 | 10 | 4463 | |||
XM_017005189.1 | XP_016860678.1 | 11 | 5358 | |||
XM_017005190.1 | XP_016860679.1 | 10 | 4327 | |||
XM_017005191.1 | XP_016860680.1 | 10 | 4406 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 5293 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 7310 | |||
XM_017005194.1 | XP_016860683.1 | 10 | 5166 | |||
XM_017005195.1 | XP_016860684.1 | 11 | 4504 | |||
XM_017005196.1 | XP_016860685.1 | 8 | 4129 |
Gene Symbol: HGSNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057293.1 | - | 1 | 216 |
Gene Symbol: LOC400002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135047.1 | 1 | 554 | |||
GenBank mRNA | AK128230.1 | - | 1 | 555 |
Gene Symbol: METTL21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127395.2 | NP_001120867.1 | 4 | 3598 | ||
NM_145280.5 | NP_660323.3 | 4 | 3459 |
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126330.1 | - | 9 | 3248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
