siRNA Details
Gene Symbol: MAPK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010302.1 | XP_016865791.1 | 10 | 2626 |
Gene Symbol: LOC55338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130914.1 | 3 | 1803 | |||
GenBank mRNA | AK002104.1 | - | 2 | 2497 | ||
AK002154.1 | - | 2 | 1724 | |||
AK023417.1 | - | 3 | 1804 | |||
AK024354.1 | - | 1 | 1450 |
Gene Symbol: TFPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209866.1 | - | 1 | 776 |
Gene Symbol: ZNF471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321768.1 | NP_001308697.1 | 4 | 3487 | ||
NM_020813.3 | NP_065864.2 | 5 | 3614 | |||
XM_011527148.1 | XP_011525450.1 | 5 | 6441 | |||
XM_017027032.1 | XP_016882521.1 | 4 | 6314 | |||
GenBank mRNA | AB037817.1 | - | 2 | 3256 | ||
AL831845.1 | - | 5 | 3614 | |||
BC016830.1 | - | 1 | 88 | |||
BC028227.1 | - | 1 | 230 | |||
BC125221.1 | - | 5 | 3572 | 40124078 | ||
BC125222.1 | - | 5 | 3572 | 40124080 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
