siRNA Details
Gene Symbol: RNF208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031297.4 | NP_112587.2 | 1 | 658 | ||
GenBank mRNA | AF416715.1 | - | 1 | 339 | ||
AL136540.1 | - | 1 | 341 | |||
BC016958.1 | - | 1 | 347 | 4151712 | ||
DN995061.1 | - | 1 | 421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
