siRNA Details
Gene Symbol: GATSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145064.2 | NP_001138536.1 | 3 | 385 | ||
XM_017012574.1 | XP_016868063.1 | 3 | 510 | |||
GenBank mRNA | BC147030.1 | - | 3 | 220 | 9056958 | |
BC147031.1 | - | 3 | 220 | 9056959 | ||
BC150658.1 | - | 3 | 385 | 9057029 | ||
DN831815.1 | - | 3 | 144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
