siRNA Details
Gene Symbol: GATSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145064.2 | NP_001138536.1 | 8 | 1078 | ||
GenBank mRNA | BC147030.1 | - | 8 | 913 | 9056958 | |
BC147031.1 | - | 8 | 913 | 9056959 | ||
BC150658.1 | - | 8 | 1078 | 9057029 | ||
DR731193.1 | - | 4 | 397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
