siRNA Details
Gene Symbol: RPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001028.2 | NP_001019.1 | 3 | 213 | ||
GenBank mRNA | AK311883.1 | - | 3 | 213 | ||
BC004294.1 | - | 1 | 241 | 3622031 | ||
BC004986.1 | - | 3 | 196 | 2905996 | ||
M64716.1 | - | 3 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
