siRNA Details
Gene Symbol: RPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001028.2 | NP_001019.1 | 4 | 375 | ||
GenBank mRNA | AK311883.1 | - | 4 | 375 | ||
BC004294.1 | - | 1 | 403 | 3622031 | ||
BC004986.1 | - | 4 | 358 | 2905996 | ||
M64716.1 | - | 4 | 383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
