siRNA Details
Gene Symbol: RPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256580.2 | NP_001243509.2 | 5, 6 | 744 | ||
NM_001303624.1 | NP_001290553.1 | 5, 6 | 647 | |||
NM_001303625.1 | NP_001290554.1 | 6, 7 | 856 | |||
NM_001303626.1 | NP_001290555.1 | 5, 6 | 511 | |||
NM_006013.4 | NP_006004.3 | 6, 7 | 852 | |||
GenBank mRNA | AF486812.1 | - | 1 | 521 | ||
AK026568.1 | - | 5, 6 | 644 | |||
AK130582.1 | - | 6, 7 | 540 | |||
BC003358.1 | - | 5, 6 | 523 | 2900175 | ||
BC026276.1 | - | 1 | 540 | 4276627 | ||
BC071918.1 | - | 1 | 518 | 6731213 | ||
CA776692.1 | - | 4, 5 | 473 | |||
CR456797.1 | - | 5, 6 | 499 | |||
CR542069.1 | - | 5, 6 | 499 | |||
DQ369703.1 | - | 5, 6 | 499 | |||
DQ369704.1 | - | 5, 6 | 499 | |||
DQ369705.1 | - | 5, 6 | 499 | |||
DQ369706.1 | - | 5, 6 | 499 | |||
DQ369707.1 | - | 5, 6 | 499 | |||
DQ369708.1 | - | 5, 6 | 499 | |||
DQ369709.1 | - | 5, 6 | 499 | |||
DQ369710.1 | - | 5, 6 | 499 | |||
DQ369711.1 | - | 5, 6 | 499 | |||
DQ369712.1 | - | 5, 6 | 499 | |||
DQ369713.1 | - | 5, 6 | 499 | |||
DQ369714.1 | - | 5, 6 | 499 | |||
DQ369715.1 | - | 5, 6 | 499 | |||
DQ369716.1 | - | 5, 6 | 499 | |||
KJ534934.1 | - | 5, 6 | 499 | |||
KJ534935.1 | - | 3, 4 | 205 | |||
M64241.1 | - | 6, 7 | 540 | |||
M73791.1 | - | 5, 6 | 513 | |||
S35960.1 | - | 1 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
