siRNA Details
Gene Symbol: GAGE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012196.1 | NP_036328.1 | 5 | 413 | ||
GenBank mRNA | AF055473.1 | - | 5 | 413 | ||
BC143620.1 | - | 5 | 422 | 9052133 |
Gene Symbol: GAGE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098413.3 | NP_001091883.3 | 5 | 457 |
Gene Symbol: GAGE12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127345.1 | NP_001120817.1 | 5 | 425 | ||
GenBank mRNA | BC130374.1 | - | 5 | 425 | 40146405 | |
BC143615.1 | - | 5 | 425 | 9052128 | ||
BC143619.1 | - | 5 | 425 | 9052132 |
Gene Symbol: GAGE12C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098408.1 | NP_001091878.1 | 5 | 460 |
Gene Symbol: GAGE12D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127199.1 | NP_001120671.1 | 5 | 471 |
Gene Symbol: GAGE12E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098418.1 | NP_001091888.1 | 5 | 460 |
Gene Symbol: GAGE12F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098405.2 | NP_001091875.1 | 5 | 494 | ||
GenBank mRNA | CF780547.1 | - | 5 | 417 | ||
HY016295.1 | - | 5 | 494 |
Gene Symbol: GAGE12G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098409.1 | NP_001091879.1 | 5 | 460 |
Gene Symbol: GAGE12H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098410.1 | NP_001091880.1 | 5 | 460 |
Gene Symbol: GAGE12I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001477.1 | NP_001468.1 | 5 | 398 |
Gene Symbol: GAGE12J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098406.2 | NP_001091876.2 | 5 | 467 | ||
GenBank mRNA | BG186708.1 | - | 2 | 143 | ||
BX108227.1 | - | 5 | 408 |
Gene Symbol: GAGE13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098412.2 | NP_001091882.2 | 5 | 439 |
Gene Symbol: GAGE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127212.1 | NP_001120684.1 | 5 | 438 | ||
GenBank mRNA | BC069309.1 | - | 5 | 413 | 7262139 | |
BC069397.1 | - | 5 | 413 | 7262151 | ||
BC069558.1 | - | 5 | 391 | 7262092 | ||
BG184057.1 | - | 2 | 138 |
Gene Symbol: GAGE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098411.3 | NP_001091881.1 | 5 | 424 |
Gene Symbol: GAGE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001472.2 | NP_001463.1 | 5 | 424 | ||
GenBank mRNA | BC113926.1 | - | 5 | 395 | 40021787 | |
U19143.1 | - | 5 | 424 |
Gene Symbol: GAGE2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098407.1 | NP_001091877.1 | 5 | 457 |
Gene Symbol: GAGE2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127200.2 | NP_001120672.2 | 5 | 457 | ||
GenBank mRNA | BC018052.1 | - | 5 | 437 | 4812462 |
Gene Symbol: GAGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U19144.1 | - | 446 |
Gene Symbol: GAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001474.1 | NP_001465.1 | 5 | 426 | ||
GenBank mRNA | BC171769.1 | - | 5 | 392 | 9054423 | |
U19145.1 | - | 5 | 426 |
Gene Symbol: GAGE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001475.1 | NP_001466.1 | 5 | 418 | ||
GenBank mRNA | BC024914.1 | - | 5 | 431 | 4471969 | |
U19146.1 | - | 5 | 418 |
Gene Symbol: GAGE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001476.1 | NP_001467.1 | 5 | 425 | ||
GenBank mRNA | U19147.1 | - | 5 | 425 |
Gene Symbol: GAGE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021123.2 | NP_066946.1 | 5 | 397 | ||
GenBank mRNA | AF055474.1 | - | 5 | 398 | ||
AF058988.1 | - | 5 | 423 | |||
BC031628.1 | - | 5 | 416 | 5168892 | ||
BC081536.1 | - | 4 | 289 | 30524801 | ||
BC104951.1 | - | 5 | 425 | 8143954 | ||
BC121182.1 | - | 5 | 404 | 40123253 | ||
BC121183.1 | - | 5 | 404 | 40123261 | ||
BC144193.1 | - | 5 | 392 | 9052713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
