siRNA Details
Gene Symbol: RPSAP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003662.3 | 4 | 467 |
Gene Symbol: RPSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304288.1 | NP_001291217.1 | 2 | 153 | ||
NM_002295.5 | NP_002286.2 | 2 | 153 | |||
GenBank mRNA | AF503367.1 | - | 1 | 113 | ||
BC005391.1 | - | 1 | 125 | 3997019 | ||
BC008867.2 | - | 1 | 74 | 4130936 | ||
BC010418.2 | - | 1 | 105 | 4079845 | ||
BC013827.2 | - | 1 | 109 | 3446816 | ||
BC018867.2 | - | 1 | 99 | |||
BC034537.1 | - | 1 | 106 | 3455545 | ||
BC050688.1 | - | 1 | 108 | 4443949 | ||
BC053370.1 | - | 1 | 106 | 5108337 | ||
BC062714.1 | - | 1 | 129 | 3686924 | ||
BC066941.1 | - | 2 | 122 | 4820432 | ||
BC068062.1 | - | 1 | 87 | 5427114 | ||
BC070263.1 | - | 2 | 153 | 3996864 | ||
BC071693.1 | - | 1 | 115 | 3884457 | ||
BC071968.1 | - | 1 | 106 | 5814758 | ||
BC071969.1 | - | 1 | 94 | 6483856 | ||
BC071970.1 | - | 1 | 79 | 6484091 | ||
BC073863.1 | - | 1 | 91 | 3924066 | ||
BC107567.1 | - | 1 | 126 | 3681016 | ||
BT007219.1 | - | 1 | 44 | |||
BU932677.1 | - | 83 | ||||
J03799.1 | - | 1 | 81 | |||
S37431.1 | - | 1 | 59 | |||
X15005.1 | - | 2 | 107 | |||
X61156.1 | - | 1 | 14 |
Gene Symbol: RPSAP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF284768.1 | - | 1 | 121 |
Gene Symbol: RPSAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026890.1 | 1 | 144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
