siRNA Details
Gene Symbol: RPSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304288.1 | NP_001291217.1 | 4 | 490 | ||
NM_002295.5 | NP_002286.2 | 4 | 475 | |||
GenBank mRNA | AF503367.1 | - | 1 | 435 | ||
AK055991.1 | - | 1 | 609 | |||
BC005391.1 | - | 1 | 447 | 3997019 | ||
BC008867.2 | - | 1 | 396 | 4130936 | ||
BC010418.2 | - | 1 | 427 | 4079845 | ||
BC013827.2 | - | 1 | 431 | 3446816 | ||
BC018867.2 | - | 1 | 421 | |||
BC034537.1 | - | 1 | 428 | 3455545 | ||
BC050688.1 | - | 1 | 430 | 4443949 | ||
BC053370.1 | - | 1 | 428 | 5108337 | ||
BC062714.1 | - | 1 | 451 | 3686924 | ||
BC066941.1 | - | 4 | 444 | 4820432 | ||
BC068062.1 | - | 1 | 409 | 5427114 | ||
BC070263.1 | - | 4 | 475 | 3996864 | ||
BC071693.1 | - | 1 | 437 | 3884457 | ||
BC071968.1 | - | 1 | 428 | 5814758 | ||
BC071969.1 | - | 1 | 416 | 6483856 | ||
BC071970.1 | - | 1 | 401 | 6484091 | ||
BC073863.1 | - | 1 | 413 | 3924066 | ||
BC107567.1 | - | 1 | 448 | 3681016 | ||
BT007219.1 | - | 3 | 366 | |||
J03799.1 | - | 3 | 403 | |||
S37431.1 | - | 3 | 381 | |||
X15005.1 | - | 4 | 444 | |||
X61156.1 | - | 3 | 336 |
Gene Symbol: RPSAP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003662.3 | 4 | 789 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
