siRNA Details
Gene Symbol: RPL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000985.4 | NP_000976.1 | 7 | 894 | ||
NM_001035006.2 | NP_001030178.1 | 7 | 645 | |||
NM_001199340.1 | NP_001186269.1 | 8 | 743 | |||
NM_001199341.1 | NP_001186270.1 | 7 | 744 | |||
NM_001199342.1 | NP_001186271.1 | 8 | 842 | |||
NM_001199343.1 | NP_001186272.1 | 8 | 825 | |||
NM_001199344.1 | NP_001186273.1 | 7 | 727 | |||
NM_001199345.1 | NP_001186274.1 | 6 | 604 | |||
GenBank mRNA | AK304659.1 | - | 6 | 518 | ||
AK311828.1 | - | 7 | 559 | |||
BC000502.2 | - | 6 | 797 | 2821464 | ||
BC017831.1 | - | 1 | 563 | 4251433 | ||
BC066323.1 | - | 1 | 559 | 4797583 | ||
BC066324.1 | - | 1 | 563 | 4799029 | ||
BC106031.1 | - | 1 | 530 | 4540700 | ||
BQ882430.1 | - | 7 | 709 | |||
CD387752.1 | - | 7 | 750 | |||
X53777.1 | - | 6 | 662 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001002239, BC054424, BC106165, BC003896, BC092091, and others.
Gene Ontology Categories:
