siRNA Details
Gene Symbol: RPL17-C18orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199355.1 | NP_001186284.1 | 2 | 83 | ||
GenBank mRNA | BX393840.2 | - | 2 | 85 |
Gene Symbol: RPL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000985.4 | NP_000976.1 | 3 | 440 | ||
NM_001035006.2 | NP_001030178.1 | 3 | 191 | |||
NM_001199340.1 | NP_001186269.1 | 4 | 289 | |||
NM_001199341.1 | NP_001186270.1 | 3 | 290 | |||
NM_001199342.1 | NP_001186271.1 | 4 | 388 | |||
NM_001199343.1 | NP_001186272.1 | 4 | 371 | |||
NM_001199344.1 | NP_001186273.1 | 3 | 273 | |||
GenBank mRNA | AK311720.1 | - | 2 | 364 | ||
AK311828.1 | - | 3 | 105 | |||
BC000502.2 | - | 2 | 343 | 2821464 | ||
BC017831.1 | - | 1 | 109 | 4251433 | ||
BC066323.1 | - | 1 | 105 | 4797583 | ||
BC066324.1 | - | 1 | 109 | 4799029 | ||
BC106031.1 | - | 1 | 76 | 4540700 | ||
BF670086.1 | - | 287 | ||||
BG497914.1 | - | 3 | 356 | |||
BG758380.1 | - | 4 | 307 | |||
BG827384.1 | - | 3 | 191 | |||
BI086293.1 | - | 3 | 162 | |||
BQ882430.1 | - | 3 | 256 | |||
CB178432.1 | - | 2 | 356 | |||
CD386294.1 | - | 3 | 210 | |||
CD387752.1 | - | 3 | 295 | |||
X53777.1 | - | 2 | 208 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
