siRNA Details
Gene Symbol: RPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000976.3 | NP_000967.1 | 5 | 434 | ||
GenBank mRNA | AF037643.1 | - | 5 | 404 | ||
AK026526.1 | - | 1 | 407 | |||
AK057602.1 | - | 3 | 1905 | |||
BC050644.1 | - | 1 | 387 | 5108312 | ||
BC059950.1 | - | 2 | 276 | 3454522 | ||
BC094831.1 | - | 1 | 382 | 5473344 | ||
BC105603.1 | - | 1 | 411 | 6698283 | ||
CB155565.1 | - | 5 | 434 | |||
L06505.1 | - | 5 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
