siRNA Details
Gene Symbol: HSD3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000198.3 | NP_000189.1 | 3 | 452 | ||
NM_001166120.1 | NP_001159592.1 | 3 | 529 | |||
GenBank mRNA | AK222997.1 | - | 3 | 450 | ||
AK313413.1 | - | 3 | 422 | |||
BC038419.1 | - | 3 | 427 | 5167092 | ||
BC131488.1 | - | 2 | 1054 | 40037863 | ||
BP197992.1 | - | 3 | 558 | |||
CF596786.1 | - | 3 | 426 | |||
M67466.1 | - | 3 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
