siRNA Details
Gene Symbol: SMIM11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058182.4 | NP_478062.1 | 3 | 352 | ||
XM_017028397.1 | XP_016883886.1 | 3 | 311 | |||
XM_017028398.1 | XP_016883887.1 | 3 | 311 | |||
XM_017028399.1 | XP_016883888.1 | 3 | 860 | |||
GenBank mRNA | AK057880.1 | - | 3 | 528 | ||
AK092693.1 | - | 3 | 287 | |||
AY033902.1 | - | 2 | 234 | |||
AY081144.1 | - | 3 | 302 | |||
BC015596.2 | - | 3 | 283 | 4647871 |
Gene Symbol: SMIM11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313692.1 | NP_001300621.1 | 3 | 322 | ||
XM_017028203.1 | XP_016883692.1 | 3 | 311 | |||
XM_017028204.1 | XP_016883693.1 | 3 | 311 | |||
XM_017028205.1 | XP_016883694.1 | 3 | 860 | |||
GenBank mRNA | AW901268.1 | - | 3 | 289 | ||
BP194116.1 | - | 3 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
