siRNA Details
Gene Symbol: RPS27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135592.2 | NP_001129064.1 | 4 | 348 | ||
NM_001177413.1 | NP_001170884.1 | 3 | 272 | |||
NM_002954.5 | NP_002945.1 | 4 | 446 | |||
GenBank mRNA | AK093614.1 | - | 1 | 1453 | ||
AK225079.1 | - | 1 | 163 | |||
AK225718.1 | - | 1 | 163 | |||
AK315695.1 | - | 4 | 163 | |||
AY952889.1 | - | 1 | 150 | |||
BC001392.2 | - | 1 | 152 | 3050927 | ||
BC042362.1 | - | 4 | 163 | 4831143 | ||
BC053371.1 | - | 1 | 152 | 5531043 | ||
BC066293.1 | - | 1 | 163 | 30336950 | ||
BF678026.1 | - | 3 | 271 | |||
BQ276758.1 | - | 3 | 166 | |||
BQ421613.1 | - | 1 | 31 | |||
CK004647.1 | - | 4 | 322 | |||
CN344335.1 | - | 3 | 147 | |||
CR746519.1 | - | 4 | 457 | |||
DA527860.1 | - | 3 | 248 | |||
DB505702.1 | - | 4 | 348 | |||
EU794671.1 | - | 4 | 163 | |||
M10939.1 | - | 3 | 113 | |||
S79522.1 | - | 4 | 163 | |||
X63237.1 | - | 1 | 150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
