siRNA Details
Gene Symbol: RPS27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135592.2 | NP_001129064.1 | 5 | 512 | ||
NM_001177413.1 | NP_001170884.1 | 4 | 436 | |||
NM_002954.5 | NP_002945.1 | 5 | 610 | |||
GenBank mRNA | AK225079.1 | - | 1 | 327 | ||
AK225718.1 | - | 1 | 327 | |||
AK315695.1 | - | 5 | 327 | |||
AY952889.1 | - | 1 | 314 | |||
BC001392.2 | - | 1 | 316 | 3050927 | ||
BC042362.1 | - | 4 | 952 | 4831143 | ||
BC053371.1 | - | 1 | 316 | 5531043 | ||
BC066293.1 | - | 1 | 327 | 30336950 | ||
BF678026.1 | - | 4 | 436 | |||
BQ276758.1 | - | 4 | 330 | |||
BQ421613.1 | - | 1 | 195 | |||
CK004647.1 | - | 5 | 486 | |||
CN344335.1 | - | 4 | 311 | |||
DA527860.1 | - | 4 | 412 | |||
EU794671.1 | - | 5 | 327 | |||
M10939.1 | - | 4 | 277 | |||
S79522.1 | - | 5 | 327 | |||
X63237.1 | - | 1 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
