siRNA Details
Gene Symbol: RPS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022551.2 | NP_072045.1 | 3 | 179 | ||
GenBank mRNA | AY536376.1 | - | 4 | 316 | ||
BC101786.1 | - | 3 | 174 | 8069292 | ||
BC101788.1 | - | 3 | 174 | 8069294 | ||
BC106063.1 | - | 3 | 232 | 4386992 | ||
X69150.1 | - | 3 | 180 |
Gene Symbol: RPS18P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077247.1 | 1 | 603 | |||
GenBank mRNA | BC071678.1 | - | 1 | 247 | ||
EB387363.1 | - | 1 | 603 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011296, BC091729, AK159437, M76763, AK146354, and others.
Gene Ontology Categories:
