siRNA Details
Gene Symbol: NDUFA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002489.3 | NP_002480.1 | 2 | 222 | ||
GenBank mRNA | AF201077.1 | - | 1 | 146 | ||
AK024837.1 | - | 2 | 171 | |||
AK130950.1 | - | 2 | 162 | |||
BC101794.1 | - | 2 | 146 | 8069300 | ||
BC101796.1 | - | 2 | 146 | 8069302 | ||
BC105295.1 | - | 2 | 193 | 6739636 | ||
BG623521.1 | - | 223 | ||||
CR407618.1 | - | 2 | 93 | |||
CR541716.1 | - | 2 | 93 | |||
U94586.1 | - | 2 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
