siRNA Details
Gene Symbol: KATNAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014380.2 | NP_001014402.1 | 11 | 4080 | ||
NM_032116.4 | NP_115492.1 | 11 | 4122 | |||
XM_005266574.2 | XP_005266631.1 | 11 | 4264 | |||
XM_017020791.1 | XP_016876280.1 | 10 | 3966 | |||
XM_017020792.1 | XP_016876281.1 | 10 | 3987 | |||
XM_017020793.1 | XP_016876282.1 | 8 | 4296 | |||
GenBank mRNA | BX647147.1 | - | 1 | 825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
