siRNA Details
Gene Symbol: CFHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002113.2 | NP_002104.2 | 2 | 313 | ||
GenBank mRNA | AK290830.1 | - | 2 | 303 | ||
BC016755.1 | - | 2 | 291 | 3934474 | ||
BC107771.1 | - | 2 | 309 | 4712577 | ||
BG568702.1 | - | 2 | 312 | |||
M65292.1 | - | 2 | 276 | |||
M65293.1 | - | 2 | 274 | |||
X56209.1 | - | 2 | 191 | |||
X56210.1 | - | 2 | 239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
