siRNA Details
Gene Symbol: ZNF767P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027788.1 | 2 | 490 | |||
NR_027789.1 | 2 | 490 | ||||
GenBank mRNA | AK022762.1 | - | 2 | 427 | ||
DA249066.1 | - | 2 | 382 | |||
DA418417.1 | - | 2 | 401 | |||
DC295958.1 | - | 2 | 490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
