siRNA Details
Gene Symbol: NOMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004067.3 | NP_001004067.1 | 13 | 1609 | ||
XM_005255318.1 | XP_005255375.1 | 13 | 1603 | |||
GenBank mRNA | AU132793.1 | - | 6 | 671 | ||
BC006531.2 | - | 2 | 230 | |||
BM507095.1 | - | 4 | 477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
