siRNA Details
Gene Symbol: XCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003175.3 | NP_003166.1 | 3 | 254 | ||
GenBank mRNA | BC069360.1 | - | 3 | 221 | 7262751 | |
BC070308.1 | - | 3 | 254 | 30413788 | ||
CD521967.1 | - | 3 | 257 |
Gene Symbol: XCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U23772.1 | - | 3 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
