siRNA Details
Gene Symbol: CCZ1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198097.3 | NP_932765.1 | 15 | 1676 | ||
GenBank mRNA | AK296026.1 | - | 14 | 1482 | ||
AL050215.1 | - | 13 | 1318 | |||
BC010130.2 | - | 15 | 1611 | 3953739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
