siRNA Details
Gene Symbol: GOLGA8IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024074.1 | 2 | 221 | |||
GenBank mRNA | AK093104.1 | - | 2 | 221 |
Gene Symbol: GOLGA8M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282468.1 | NP_001269397.1 | 2 | 221 | ||
XM_005268284.4 | XP_005268341.1 | 2 | 1020 | |||
XM_005268285.4 | XP_005268342.1 | 2 | 1020 | |||
XM_006720654.3 | XP_006720717.1 | 2 | 164 | |||
XM_011521951.2 | XP_011520253.1 | 3 | 589 | |||
XM_011521952.2 | XP_011520254.1 | 2 | 1020 | |||
XM_011521953.2 | XP_011520255.1 | 2 | 1020 | |||
XM_011521954.2 | XP_011520256.1 | 2 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
