siRNA Details
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 5 | 556 | ||
NM_018172.2 | NP_060642.2 | 5 | 577 | |||
NM_152563.2 | NP_689776.1 | 4 | 475 | |||
GenBank mRNA | AK001523.1 | - | 5 | 551 | ||
AK130709.1 | - | 4 | 472 | |||
AK293221.1 | - | 4 | 591 | |||
AK311428.1 | - | 5 | 533 | |||
AK311518.1 | - | 5 | 556 | |||
BC032519.1 | - | 4 | 475 | 5494088 | ||
BQ921333.1 | - | 5 | 526 |
Gene Symbol: FAM86KP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008896.1 | XP_016864385.1 | 3 | 627 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
