siRNA Details
Gene Symbol: SLC35G5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054028.1 | NP_473369.1 | 1 | 308 | ||
GenBank mRNA | AJ291677.1 | - | 1 | 308 | ||
BC131696.1 | - | 1 | 275 | 40120574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
