siRNA Details
Gene Symbol: BRD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173984.2 | NP_001167455.1 | 12 | 1520 | ||
NM_013263.4 | NP_037395.2 | 12 | 1520 | |||
XM_011523046.2 | XP_011521348.1 | 12 | 1606 | |||
XM_011523047.2 | XP_011521349.1 | 12 | 1606 | |||
XM_011523048.2 | XP_011521350.1 | 11 | 1498 | |||
XM_011523049.2 | XP_011521351.1 | 11 | 1498 | |||
XM_011523050.2 | XP_011521352.1 | 8 | 1626 | |||
XM_017023179.1 | XP_016878668.1 | 11 | 1312 | |||
XM_017023180.1 | XP_016878669.1 | 11 | 1312 | |||
XM_017023181.1 | XP_016878670.1 | 8 | 1643 | |||
GenBank mRNA | AF152604.1 | - | 12 | 1413 | ||
AF213969.1 | - | 12 | 1365 | |||
AJ271881.1 | - | 12 | 1478 | |||
AK027308.1 | - | 5 | 491 | |||
AK074613.1 | - | 6 | 657 | |||
BC001611.1 | - | 8 | 781 | |||
BC050728.1 | - | 13 | 1427 | 6149862 | ||
BC094706.1 | - | 13 | 1397 | 30719813 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
