siRNA Details
Gene Symbol: FAM86KP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008895.1 | XP_016864384.1 | 3 | 193 | ||
XM_017008897.1 | XP_016864386.1 | 3 | 193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
