siRNA Details
Gene Symbol: GOLGA6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164404.1 | NP_001157876.1 | 9 | 731 | ||
XM_017022519.1 | XP_016878008.1 | 9 | 885 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
