siRNA Details
Gene Symbol: UBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281716.1 | NP_001268645.1 | 2 | 1015 | ||
NM_001281717.1 | NP_001268646.1 | 2 | 986 | |||
NM_001281718.1 | NP_001268647.1 | 2 | 946 | |||
NM_001281719.1 | NP_001268648.1 | 2 | 944 | |||
NM_001281720.1 | NP_001268649.1 | 2 | 790 | |||
NM_018955.3 | NP_061828.1 | 2 | 1159 | |||
GenBank mRNA | AK129498.1 | - | 2 | 861 | ||
AK300882.1 | - | 3 | 637 | |||
BC000379.2 | - | 2 | 903 | 2820408 | ||
BC009301.2 | - | 2 | 849 | 4126622 | ||
BC015127.1 | - | 2 | 865 | 3929901 | ||
BC026301.1 | - | 2 | 861 | 4280629 | ||
BC031027.1 | - | 2 | 863 | 4715377 | ||
BC038999.1 | - | 2 | 872 | 4943285 | ||
BC046123.2 | - | 2 | 851 | 3879581 | ||
GQ472200.1 | - | 2 | 903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
