siRNA Details
Gene Symbol: HLA-DRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK297032.1 | - | 3 | 417 | ||
AK304429.1 | - | 3 | 300 | |||
AK313123.1 | - | 3 | 409 | |||
BC032350.1 | - | 3 | 385 | 5214156 | ||
BC071659.1 | - | 3 | 393 | 5162925 | ||
BM849755.1 | - | 3 | 455 | |||
CR457013.1 | - | 3 | 345 | |||
J00194.1 | - | 3 | 371 | |||
K01171.1 | - | 3 | 359 | |||
M60334.1 | - | 3 | 372 | |||
V00523.1 | - | 2 | 254 |
Gene Symbol: HLA-DQA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF522250.1 | - | 365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
