siRNA Details
Gene Symbol: MAP1LC3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085481.1 | NP_001078950.1 | 2 | 492 | ||
XM_011538652.2 | XP_011536954.1 | 2 | 665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
